missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine UMOD Protein

A cDNA sequence encoding the UMOD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13890-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985029

  • 6549.36 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UMOD Protein
Research Use Only
Recombinant Protein
Canine
RSCSECHSNA TCMEDGMVTT CSCLVGFTGS GFECVDLDEC AIPGAHNCSE GSSCMNTLGS YLCTCPDGFR LTPGLGCIDV DECSEPGLSR CHALATCINN KGNYSCVCPA GYRGDGQHCE CSPGSCGPGL DCVPVGDALV CADPCQEHRI LDEYWRSTEY GAGYTCDVGL NGWYRFTGPG GVRLAETCVP VLHCNTAAPM WLNGTHPTRD QGIVNRTACA HWRGHCCLWD ASIQVKACAG GYYVYNLTET PECYLAYCTD PTSVLGTCEE CSVEEDCKSH DGMWSCQCKQ DFNVTDLFLL DRLECRPNDI KVSLSKCQLK SLGFEKVFMY LRDSQCSGFN ERGDRDWVSV VTPARDGPCG TVMVRNETHA TYSNTLYLAD EIVIRDRNIK INFECSYPLD MKVSLETSLQ PIVSSLNISV GGTGMFTVRM ALFQTPDYTQ PYQGSSVTLT TEAFLYVGTM LDGGDLSRFA LLMTNCYATP SSNATDPLKY FIIQDRCPRT TDSTIQVVEN GESPQGRFSV QMFRFAGNYD LVYLHCEVYL CDIINEKCKP TCSGTRFRSG GIIDQSRVLN LGPITRKNVQ AVVSRAASS
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Canine
Untagged
The UMOD protein was lyophilized from 0.4microm filtered solution at a concentration of 0.1 mg/ml containing deionized water.
Videos
SDS
Documents
Certificates