missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cat CSF1R / MCSF Receptor / CD115 Protein
A DNA sequence encoding the Felis catus (Cat) (Felis silvestris catus) CSF1R / MCSF Receptor / CD115, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5875-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009231.1 | |
Cat CSF1R / MCSF Receptor / CD115 Protein | |
Research Use Only | |
CSF1R | |
Recombinant Protein | |
E. coli | |
VPVIQPSGPE LVVEPGTTVT LRCVGNGSVE WDGPISPHWN LDLDPPSSIL TTNNATFQNT GTYHCTEPGN PQGGNATIHL YVKDPARPWK VLAQEVTVLE GQDALLPCLL TDPALEAGVS LVRVRGRPVL RQTNYSFSPW HGFTIHKAKF IENHVYQCSA RVDGRTVTSM GIWLKVQKDI SGPATLTLEP AELVRIQGEA AQIVCSASNI DVNFDVSLRH GDTKLTISQQ SDFHDNRYQK VLTLNLDHVS FQDAGNYSCT ATNAWGNHSA SMVFRVVESA YLNLTSEQSL LQEVTVGEKV DLQVKVEAYP GLESFNWTYL GPFSDYQDKL DFVTIKDTYR YTSTLSLPRL KRSEAGRYSF LARNAGGQNA LTFELTLRYP PEVRVTMTLI NGSDTLLCEA SGYPQPSVTW VQCRSHTDRC DESAGLVLED SHSEVLSQVP FHEVIVHSLL AIGTLEHNRT YECRAFNSVG NSSQTFWPIS IGAHTQLPDE LLFTP | |
Greater than 90% as determined by SDS-PAGE. |
493706 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |