missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Collagen alpha-1(XII) chain Protein
A DNA sequence encoding the Gallus gallus (Chicken) Collagen alpha-1(XII) chain, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Name | Collagen alpha-1(XII) chain Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | COL12A1 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948229
|
enQuireBio™
QP5851-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988229
|
enQuireBio™
QP5851-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958229
|
enQuireBio™
QP5851-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978229
|
enQuireBio™
QP5851-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998229
|
enQuireBio™
QP5851-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968229
|
enQuireBio™
QP5851-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Collagen alpha-1(XII) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
COL12A1 | |
Recombinant Protein | |
Yeast | |
DLVFLVDGSW SVGRNNFRYI LDFMVALVSA FDIGEEKTRV GVVQYSSDTR TEFNLNQYFR RSDLLDAIKR IPYKGGNTMT GEAIDYLVKN TFTESAGARK GFPKVAIVIT DGKAQDEVEI PARELRNIGV EVFSLGIKAA DAKELKLIAS QPSLKHVFNV ANFDGIVDIQ NEI | |
Greater than 90% as determined by SDS-PAGE. |