missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken GAPDH Protein
A DNA sequence encoding the Gallus gallus (Chicken) GAPDH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6069-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_989636.1 | |
GAPDH Protein | |
Research Use Only | |
GAPDH | |
Recombinant Protein | |
E. coli | |
VKVGVNGFGR IGRLVTRAAV LSGKVQVVAI NDPFIDLNYM VYMFKYDSTH GHFKGTVKAE NGKLVINGHA ITIFQERDPS NIKWADAGAE YVVESTGVFT TMEKAGAHLK GGAKRVIISA PSADAPMFVM GVNHEKYDKS LKIVSNASCT TNCLAPLAKV IHDNFGIVEG LMTTVHAITA TQKTVDGPSG KLWRDGRGAA QNIIPASTGA AKAVGKVIPE LNGKLTGMAF RVPTPNVSVV DLTCRLEKPA KYDDIKRVVK AAADGPLKGI LGYTEDQVVS CDFNGDSHSS TFDAGAGIAL NDHFVKLVSW YDNEFGYSNR VVDLMVHMAS KE | |
Greater than 90% as determined by SDS-PAGE. |
374193 | |
100 μg | |
Not determined. | |
Full Length | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |