missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia HSP70 Protein
A cDNA sequence encoding the Chlamydia HSP70 (Amino Acids 462-503) was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | HSP70 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chlamydia |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15977178
|
enQuireBio™
QP11411-EC-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997178
|
enQuireBio™
QP11411-EC-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987178
|
enQuireBio™
QP11411-EC-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSP70 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Chlamydia HSP70 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 8M Urea, 60 mM NaCl and 50% glycerol. |