missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant CMV CMV Pp150 Protein
A cDNA sequence encoding the CMV Pp150 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11462-500ug
Additional Details : Weight : 0.01000kg
Specifications
Pp150 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 25mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
CMV | |
Untagged | |
CMV Pp150 protein is >95% pure as determined by 10% PAGE (coomassie staining). |