missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.
437.00 € - 1245.00 €
Specifications
Name | Uncharacterized protein UL131 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | UL131 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047340
|
enQuireBio™
QP7086-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067340
|
enQuireBio™
QP7086-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057340
|
enQuireBio™
QP7086-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Uncharacterized protein UL131 Protein | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
UL131 | |
Recombinant Protein | |
E. coli | |
MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR | |
Greater than 80% as determined by SDS-PAGE. |