missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 Protein
A cDNA sequence encoding the Dengue Envelope-1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11619-10ug
Additional Details : Weight : 0.01000kg
Specifications
Dengue Envelope-1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
mrcvgignrd fveglsgatw vdvvlehgsc vttmakdkpt ldiellktev tnpavlrklc ieakisnttt dsrcptqgea tlveeqdanf vcrrtfvdrg wgngcglfgk gslltcakfk cvtklegkiv qyenlkysvi vtvhtgdqhq vgnettehgt iatitpqapt seiqltdyga ltldcsprtg ldfnemvllt mkekswlvhk qwfldlplpw tsgastsqet wnrqdllvtf ktahakkqea vvlgsqegam htaltgatei qtsgtttifa ghlkcrlkmd kltlkgisyv mctgsfklek evaetqhgtv lvqvkyegtd apckipfssq dekgvtqngr lvtanpivtd kekpvnieae ppfgesyivv gagekalkls wfkkg | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Dengue Envelope-1 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |