missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus, ST1 Dengue NS1 Protein

A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10495-100 ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15904048

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Dengue Virus, ST1 Dengue NS1 Protein
Research Use Only
Recombinant Protein
E. coli
Phosphate buffer and 25mM arginine.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus, ST1
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers