missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Tight junction protein ZO-1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Tight junction protein ZO-1, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_001003140.1 |
---|---|
Gene ID (Entrez) | 403752 |
Name | Tight junction protein ZO-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035930
|
enQuireBio™
QP6798-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055930
|
enQuireBio™
QP6798-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045930
|
enQuireBio™
QP6798-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065930
|
enQuireBio™
QP6798-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075930
|
enQuireBio™
QP6798-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085930
|
enQuireBio™
QP6798-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003140.1 | |
Tight junction protein ZO-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |
403752 | |
Research Use Only | |
TJP1 | |
Recombinant Protein | |
E. coli | |
VVATARGVFN NNGGVLSSIE TGVSIIIPQG AIPEGVEQEI YFKVCRDNSI LPPLDKEKGE TLLSPLVMCG PHGLKFLKPV ELRLPHCASM TPDGWSFALK SSDSSSGDPK TWQNKCLPGD PNYLVGANCV SVLIDHF | |
Greater than 90% as determined by SDS-PAGE. |