missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S2, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | WP_000246882.1 |
---|---|
Name | 30S ribosomal protein S2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | rpsB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039880
|
enQuireBio™
QP7628-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059880
|
enQuireBio™
QP7628-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049880
|
enQuireBio™
QP7628-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069880
|
enQuireBio™
QP7628-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079880
|
enQuireBio™
QP7628-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089880
|
enQuireBio™
QP7628-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000246882.1 | |
Research Use Only | |
rpsB | |
Recombinant Protein | |
E. coli | |
MATVSMRDML KAGVHFGHQT RYWNPKMKPF IFGARNKVHI INLEKTVPMF NEALAELNKI ASRKGKILFV GTKRAASEAV KDAALSCDQF FVNHRWLGGM LTNWKTVRQS IKRLKDLETQ SQDGTFDKLT KKEALMRTRE LEKLENSLGG IKDMGGLPDA LFVIDADHEH IAIKEANNLG IPVFAIVDTN SDPDGVDFVI PGNDDAIRAV TLYLGAVAAT VREGRSQDLA SQAEESFVEA E | |
Greater than 90% as determined by SDS-PAGE. |
30S ribosomal protein S2 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |