missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli ACKA Protein
A cDNA sequence encoding the ACKA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10939-20ug
Additional Details : Weight : 0.01000kg
Specifications
ACKA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSKLVL VLNCGSSSLK FAIIDAVNGE EYLSGLAECF HLPEARIKWK MDGNKQEAAL GAGAAHSEAL NFIVNTILAQ KPELSAQLTA IGHRIVHGGE KYTSSVVIDE SVIQGIKDAA SFAPLHNPAH LIGIEEALKS FPQLKDKNVA VFDTAFHQTM PEESYLYALP YNLYKEHGIR RYGAHGTSHF YVTQEAAKML NKPVEELNII TCHLGNGGSV SAIRNGKCVD TSMGLTPLEG LVMGTRSGDI DPAIIFHLHD TLGMSVDAIN KLLTKESGLL GLTEVTSDCR YVEDNYATKE DAKRAMDVYC HRLAKYIGAY TALMDGRLDA VVFTGGIGEN AAMVRELSLG KLGVLGFEVD HERNLAARFG KSGFINKEGT RPAVVIPTNE ELVIAQDASR LTA | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
ACKA protein (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |