missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein
A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7131-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_001033658.1 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
CFA / I fimbrial subunit E Protein | |
Research Use Only | |
cfaE | |
Recombinant Protein | |
Yeast | |
MNKILFIFTL FFSSGFFTFA VSADKNPGSE NMTNTIGPHD RGGSSPIYNI LNSYLTAYNG SHHLYDRMSF LCLSSQNTLN GACPSSDAPG TATIDGETNI TLQFTEKRSL IKRELQIKGY KQFLFKNANC PSKLALNSSH FQCNREQASG ATLSLYIPAG ELNKLPFGGV WNAVLKLNVK RRYDTTYGTY TINITVNLTD KGNIQIWLPQ FKSNARVDLN LRPTGGGTYI GRNSVDMCFY DGYSTNSSSL EIRFQDDNSK SDGKFYLKKI NDDSKELVYT LSLLLAGKNL TPTNGQALNI NTASLETNWN RITAVTMPEI SVPVLCWPGR LQLDAKVKNP EAGQYMGNIK ITFTPSSQTL | |
Greater than 80% as determined by SDS-PAGE. |