missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Elongation factor P Protein

A DNA sequence encoding the Escherichia coli (strain K12) Elongation factor P, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7316-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_418571.1
Elongation factor P Protein
Research Use Only
efp
Recombinant Protein
E. coli
ATYYSNDFRA GLKIMLDGEP YAVEASEFVK PGKGQAFARV KLRRLLTGTR VEKTFKSTDS AEGADVVDMN LTYLYNDGEF WHFMNNETFE QLSADAKAIG DNAKWLLDQA ECIVTLWNGQ PISVTPPNFV ELEIVDTDPG LKGDTAGTGG KPATLSTGAV VKVPLFVQIG EVIKVDTRSG EYVSRVK
Greater than 90% as determined by SDS-PAGE.
948661
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers