missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ferric uptake regulation Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ferric uptake regulation protein, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_415209.1 |
---|---|
Gene ID (Entrez) | 945295 |
Name | Ferric uptake regulation Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008650
|
enQuireBio™
QP7394-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048650
|
enQuireBio™
QP7394-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018650
|
enQuireBio™
QP7394-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038650
|
enQuireBio™
QP7394-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058650
|
enQuireBio™
QP7394-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028650
|
enQuireBio™
QP7394-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415209.1 | |
Ferric uptake regulation Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945295 | |
Research Use Only | |
fur | |
Recombinant Protein | |
Yeast | |
TDNNTALKKA GLKVTLPRLK ILEVLQEPDN HHVSAEDLYK RLIDMGEEIG LATVYRVLNQ FDDAGIVTRH NFEGGKSVFE LTQQHHHDHL ICLDCGKVIE FSDDSIEARQ REIAAKHGIR LTNHSLYLYG HCAEGDCRED EHAHEGK | |
Greater than 90% as determined by SDS-PAGE. |