missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli fkpA Protein
A DNA sequence encoding the Escherichia coli (strain K12) fkpA, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | WP_000838261.1 |
---|---|
Name | fkpA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | fkpA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017920
|
enQuireBio™
QP7261-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037920
|
enQuireBio™
QP7261-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027920
|
enQuireBio™
QP7261-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047920
|
enQuireBio™
QP7261-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057920
|
enQuireBio™
QP7261-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067920
|
enQuireBio™
QP7261-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000838261.1 | |
Research Use Only | |
fkpA | |
Recombinant Protein | |
E. coli | |
AEAAKPATTA DSKAAFKNDD QKSAYALGAS LGRYMENSLK EQEKLGIKLD KDQLIAGVQD AFADKSKLSD QEIEQTLQAF EARVKSSAQA KMEKDAADNE AKGKEYREKF AKEKGVKTSS TGLVYQVVEA GKGEAPKDSD TVVVNYKGTL IDGKEFDNSY TRGEPLSFRL DGVIPGWTEG LKNIKKGGKI KLVIPPELAY GKAGVPGIPP NSTLVFDVEL LDVKPAPKAD AKPEADAKAA DSAKK | |
Greater than 90% as determined by SDS-PAGE. |
fkpA Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |