missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli insE1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) insE1, was expressed in the hosts and tags indicated.
665.00 € - 1850.00 €
Specifications
Accession Number | NP_061380.1 |
---|---|
Gene ID (Entrez) | 1263530 |
Name | insE1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932878
|
enQuireBio™
QP10112-MA-20UG |
20 μg |
665.00 €
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922878
|
enQuireBio™
QP10112-MA-100UG |
100 μg |
1850.00 €
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942878
|
enQuireBio™
QP10112-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_061380.1 | |
insE1 Protein | |
Not determined. | |
Full Length | |
E. coli | |
Flag | |
Tris-based buffer, 50% glycerol |
1263530 | |
Research Use Only | |
insE1 | |
Recombinant Protein | |
Mammal | |
MTKTVSTSKK PRKQHSPEFR SEALKLAERI GVTAAARELS LYESQLYNWR SKQQNQQTSS ERELEMSTEI ARLKRQLAER DEELAILQKA ATYFAKRLK | |
Greater than 90% as determined by SDS-PAGE. |