missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli LACTB Protein
A cDNA sequence encoding the LACTB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10738-2ug
Additional Details : Weight : 0.01000kg
Specifications
948669 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is >700 units/mg, in which One unit will hydrolyze 1.0umole of Nitrocefin per minute at pH 7.0 at 37C. | |
E. coli | |
His | |
The LACTB solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
LACTB Protein | |
Research Use Only | |
ampC | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAPQQINDIV HRTITPLIEQ QKIPGMAVAV IYQGKPYYFT WGYADIAKKQ PVTQQTLFEL GSVSKTFTGV LGGDAIARGE IKLSDPTTKY WPELTAKQWN GITLLHLATY TAGGLPLQVP DEVKSSSDLL RFYQNWQPAW APGTQRLYAN SSIGLFGALA VKPSGLSFEQ AMQTRVFQPL KLNHTWINVP PAEEKNYAWG YREGKAVHVS PGALDAEAYG VKSTIEDMAR WVQSNLKPLD INEKTLQQGI QLAQSRYWQT GDMYQGLGWE MLDWPVNPDS IINGSDNKIA LAARPVKAIT PPTPAVRASW VHKTGATGGF GSYVAFIPEK ELGIVMLANK NYPNPARVDA AWQILNALQ | |
Greater than 95.0% as determined by SDS-PAGE. |