missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli NAD(P)H-flavin reductase Protein
A DNA sequence encoding the Escherichia coli (strain K12) NAD(P)H-flavin reductase, was expressed in the hosts and tags indicated.
385.00 € - 2645.00 €
Specifications
Accession Number | NP_418286.1 |
---|---|
Gene ID (Entrez) | 948325 |
Name | NAD(P)H-flavin reductase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047261
|
enQuireBio™
QP8942-EC-10UG |
10 μg |
385.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067261
|
enQuireBio™
QP8942-EC-200UG |
200 μg |
1535.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037261
|
enQuireBio™
QP8942-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087261
|
enQuireBio™
QP8942-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077261
|
enQuireBio™
QP8942-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057261
|
enQuireBio™
QP8942-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_418286.1 | |
NAD(P)H-flavin reductase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
948325 | |
Research Use Only | |
fre | |
Recombinant Protein | |
E. coli | |
TTLSCKVTSV EAITDTVYRV RIVPDAAFSF RAGQYLMVVM DERDKRPFSM ASTPDEKGFI ELHIGASEIN LYAKAVMDRI LKDHQIVVDI PHGEAWLRDD EERPMILIAG GTGFSYARSI LLTALARNPN RDITIYWGGR EEQHLYDLCE LEALSLKHPG LQVVPVVEQP EAGWRGRTGT VLTAVLQDHG TLAEHDIYIA GRFEMAKIAR DLFCSERNAR EDRLFGDAFA FI | |
Greater than 90% as determined by SDS-PAGE. |