missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli NAD(P) transhydrogenase subunit alpha Protein
A DNA sequence encoding the Escherichia coli (strain K12) NAD(P) transhydrogenase subunit alpha, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_416120.1 |
---|---|
Gene ID (Entrez) | 946628 |
Name | NAD(P) transhydrogenase subunit alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048571
|
enQuireBio™
QP9674-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088571
|
enQuireBio™
QP9674-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058571
|
enQuireBio™
QP9674-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078571
|
enQuireBio™
QP9674-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098571
|
enQuireBio™
QP9674-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068571
|
enQuireBio™
QP9674-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_416120.1 | |
NAD(P) transhydrogenase subunit alpha Protein | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
946628 | |
Research Use Only | |
pntA | |
Recombinant Protein | |
Yeast | |
MRIGIPRERL TNETRVAATP KTVEQLLKLG FTVAVESGAG QLASFDDKAF VQAGAEIVEG NSVWQSEIIL KVNAPLDDEI ALLNPGTTLV SFIWPAQNPE LMQKLAERNV TVMAMDSVPR ISRAQSLDAL SSMANIAGYR AIVEAAHEFG RFFTGQITAA GKVPPAKVMV IGAGVAGLAA IGAANSLGAI VRAFDTRPEV KEQVQSMGAE FLELDFKEEA GSGDGYAKVM SDAFIKAEME LFAAQAKEVD IIVTTALIPG KPAPKLITRE MVDSMKAGSV IVDLAAQNGG NCEYTVPGEI FTTENGVKVI GYTDLPGRLP TQSSQLYGTN LVNLLKLLCK EKDGNITVDF DDVVIRGVTV IRAGEITWPA PPIQVSAQPQ AAQKAAPEVK TEEKCTCSPW R | |
Greater than 90% as determined by SDS-PAGE. |