missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli nrdB Protein
A DNA sequence encoding the Escherichia coli (strain K12) nrdB, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_416738.1 |
---|---|
Gene ID (Entrez) | 946732 |
Name | nrdB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026860
|
enQuireBio™
QP6953-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046860
|
enQuireBio™
QP6953-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036860
|
enQuireBio™
QP6953-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056860
|
enQuireBio™
QP6953-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066860
|
enQuireBio™
QP6953-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076860
|
enQuireBio™
QP6953-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_416738.1 | |
nrdB Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946732 | |
Research Use Only | |
nrdB | |
Recombinant Protein | |
E. coli | |
AYTTFSQTKN DQLKEPMFFG QPVNVARYDQ QKYDIFEKLI EKQLSFFWRP EEVDVSRDRI DYQALPEHEK HIFISNLKYQ TLLDSIQGRS PNVALLPLIS IPELETWVET WAFSETIHSR SYTHIIRNIV NDPSVVFDDI VTNEQIQKRA EGISSYYDEL IEMTSYWHLL GEGTHTVNGK TVTVSLRELK KKLYLCLMSV NALEAIRFYV SFACSFAFAE RELMEGNAKI IRLIARDEAL HLTGTQHMLN LLRSGADDPE MAEIAEECKQ ECYDLFVQAA QQEKDWADYL FRDGSMIGLN KDILCQYVEY ITNIRMQAVG LDLPFQTRSN PIPWINTWLV SDNVQVAPQE VEVSSYLVGQ IDSEVDTDDL SNFQL | |
Greater than 90% as determined by SDS-PAGE. |