missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) Outer-membrane lipoprotein carrier protein, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | WP_001295343.1 |
---|---|
Name | Outer-membrane lipoprotein carrier Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | lolA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048850
|
enQuireBio™
QP7432-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088850
|
enQuireBio™
QP7432-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058850
|
enQuireBio™
QP7432-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078850
|
enQuireBio™
QP7432-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098850
|
enQuireBio™
QP7432-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068850
|
enQuireBio™
QP7432-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_001295343.1 | |
Research Use Only | |
lolA | |
Recombinant Protein | |
Yeast | |
DAASDLKSRL DKVSSFHASF TQKVTDGSGA AVQEGQGDLW VKRPNLFNWH MTQPDESILV SDGKTLWFYN PFVEQATATW LKDATGNTPF MLIARNQSSD WQQYNIKQNG DDFVLTPKAS NGNLKQFTIN VGRDGTIHQF SAVEQDDQRS SYQLKSQQNG AVDAAKFTFT PPQGVTVDDQ RK | |
Greater than 90% as determined by SDS-PAGE. |
Outer-membrane lipoprotein carrier Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |