missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Periplasmic serine endoprotease DegP Protein
A DNA sequence encoding the Escherichia coli (strain K12) Periplasmic serine endoprotease DegP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7007-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_414703.1 | |
Periplasmic serine endoprotease DegP Protein | |
Research Use Only | |
degP | |
Recombinant Protein | |
Yeast | |
AETSSATTAQ QMPSLAPMLE KVMPSVVSIN VEGSTTVNTP RMPRNFQQFF GDDSPFCQEG SPFQSSPFCQ GGQGGNGGGQ QQKFMALGSG VIIDADKGYV VTNNHVVDNA TVIKVQLSDG RKFDAKMVGK DPRSDIALIQ IQNPKNLTAI KMADSDALRV GDYTVAIGNP FGLGETVTSG IVSALGRSGL NAENYENFIQ TDAAINRGNS GGALVNLNGE LIGINTAILA PDGGNIGIGF AIPSNMVKNL TSQMVEYGQV KRGELGIMGT ELNSELAKAM KVDAQRGAFV SQVLPNSSAA KAGIKAGDVI TSLNGKPISS FAALRAQVGT MPVGSKLTLG LLRDGKQVNV NLELQQSSQN QVDSSSIFNG IEGAEMSNKG KDQGVVVNNV KTGTPAAQIG LKKGDVIIGA NQQAVKNIAE LRKVLDSKPS VLALNIQRGD STIYLLMQ | |
Greater than 90% as determined by SDS-PAGE. |
947139 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |