missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ruvB Protein
A DNA sequence encoding the Escherichia coli (strain K12) ruvB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7392-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416374.1 | |
ruvB Protein | |
Research Use Only | |
ruvB | |
Recombinant Protein | |
E. coli | |
MIEADRLISA GTTLPEDVAD RAIRPKLLEE YVGQPQVRSQ MEIFIKAAKL RGDALDHLLI FGPPGLGKTT LANIVANEMG VNLRTTSGPV LEKAGDLAAM LTNLEPHDVL FIDEIHRLSP VVEEVLYPAM EDYQLDIMIG EGPAARSIKI DLPPFTLIGA TTRAGSLTSP LRDRFGIVQR LEFYQVPDLQ YIVSRSARFM GLEMSDDGAL EVARRARGTP RIANRLLRRV RDFAEVKHDG TISADIAAQA LDMLNVDAEG FDYMDRKLLL AVIDKFFGGP VGLDNLAAAI GEERETIEDV LEPYLIQQGF LQRTPRGRMA TTRAWNHFGI TPPEMP | |
Greater than 90% as determined by SDS-PAGE. |
946371 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |