missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli UPP1 Protein
A cDNA sequence encoding the UPP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13899-25ug
Additional Details : Weight : 0.01000kg
Specifications
UPP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSKSDVFHLG LTKNDLQGAT LAIVPGDPDR VEKIAALMDK PVKLASHREF TTWRAELDGK PVIVCSTGIG GPSTSIAVEE LAQLGIRTFL RIGTTGAIQP HINVGDVLVT TASVRLDGAS LHFAPLEFPA VADFECTTAL VEAAKSIGAT THVGVTASSD TFYPGQERYD TYSGRVVRHF KGSMEEWQAM GVMNYEMESA TLLTMCASQG LRAGMVAGVI VNRTQQEIPN AETMKQTESH AVKIVVEAAR RLL | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
UPP1 solution (1 mg/ml) contains 20 mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50mM NaCl and 10% glycerol. |