missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Zaire VP40 Protein
A cDNA sequence encoding the Ebola Zaire VP40 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | Ebola Zaire VP40 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Ebola Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928168
|
enQuireBio™
QP11729-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948168
|
enQuireBio™
QP11729-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938168
|
enQuireBio™
QP11729-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Ebola Zaire VP40 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
His | |
Greater than 95% as determined by 12% PAGE (Coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Zaire VP40 protein solution is supplied in Phosphate buffer, 25mM arginine and 0.02% sodium azide. |