missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ebola Virus Ebola Zaire VP40 Protein

A cDNA sequence encoding the Ebola Zaire VP40 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11729-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928168

  • 245.00 € / 100µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Ebola Zaire VP40 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Ebola Virus
His
Greater than 95% as determined by 12% PAGE (Coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Ebola Zaire VP40 protein solution is supplied in Phosphate buffer, 25mM arginine and 0.02% sodium azide.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers