missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant EBV EBNA1 Protein
A cDNA sequence encoding the EBV EBNA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11731-100ug
Additional Details : Weight : 0.01000kg
Specifications
EBNA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PBS pH 8.0 and 300mM Imidazole. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |