missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant EBV p18 Protein
A cDNA sequence encoding the EBV p18 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11733-GST-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
p18 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
GST | |
3.8 mg/ml in 50mM Tris-HCl, pH 8.0. |
Protein is >90% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
ASAGTGALAS SAPSTAVAQS ATPSVSSSIS SLRAATSGAT AAASAAAAVD TGSGGGGQPH DTAPRGARKK Q |