missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori cagA Protein
A DNA sequence encoding the Helicobacter pylori (Campylobacter pylori) cagA, was expressed in the hosts and tags indicated.
385.00 € - 2645.00 €
Specifications
Name | cagA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | cagA |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006810
|
enQuireBio™
QP6943-EC-10UG |
10 μg |
385.00 €
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096800
|
enQuireBio™
QP6943-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036810
|
enQuireBio™
QP6943-EC-500UG |
500 μg |
2115.00 €
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026810
|
enQuireBio™
QP6943-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046810
|
enQuireBio™
QP6943-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016810
|
enQuireBio™
QP6943-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
cagA Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
cagA | |
Recombinant Protein | |
E. coli | |
KVNAKIDRLN QIASGLGVVG QAAGFPLKRH DKVDDLSKVG LSRNQELAQK IDNLNQAVSE AKAGFFGNLE QTIDKLKDST KHNPMNLWVE SAKKVPASLS AKLDNYATNS HIRINSNIKN GAINEKATGM LTQKNPEWLK LVNDKIVAHN VGSVPLSEYD KIGFNQKNMK DYSDSFKFST KLNNAVKDTN SGFTQFLTNA FSTASYYCLA RENAEHGIKN VNTKGGFQKS | |
Greater than 80% as determined by SDS-PAGE. |