missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-5 Protein

A cDNA sequence encoding the HCV Core Genotype-5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12154-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959538

  • 1740.00 € / 1mg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

Hepatitis C HCV Core Genotype-5 Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, pH 8.0, 1.5M Urea, 0.2% Triton-X and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
Protein is >95% pure as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers