missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS4 Protein
A cDNA sequence encoding the HCV NS4 was constructed and used to recombinantly synthesize the protein.
373.00 € - 2175.00 €
Specifications
Name | Hepatitis C HCV NS4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909608
|
enQuireBio™
QP12176-100UG |
100 μg |
373.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929608
|
enQuireBio™
QP12176-500UG |
500 μg |
1305.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919608
|
enQuireBio™
QP12176-1MG |
1 mg |
2175.00 €
1mg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C HCV NS4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
25mM Tris-HCl pH 8.0, 1mM EDTA, 1.5M urea and 50%glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HCV NS4 Protein is >95% pure as determined by 10% PAGE (coomassie staining). |