missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-2 Protein

A cDNA sequence encoding the HCV NS5 Genotype-2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12189-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989638

Quantity Price
1 757.92 € / 500µg
5 711.76 € / 500µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Hepatitis C HCV NS5 Genotype-2 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV NS5 Genotype-2 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
25mM Tris and 1mM EDTA, 1.5M urea and 50% glycerol.
Videos
SDS
Documents
Certificates