missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-5 Protein

A cDNA sequence encoding the HCV NS5 Genotype-5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12196-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989658

Quantity Price
1 1511.71 € / 1mg
3 1421.92 € / 1mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Hepatitis C HCV NS5 Genotype-5 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV NS5 Genotype-5 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25 mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol.
Videos
SDS
Documents
Certificates