missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis protein 1
A DNA sequence encoding the Hepatitis B virus genotype D subtype ayw (isolate France/Tiollais/1979) (HBV-D) Protein X, was expressed in the hosts and tags indicated. opt
Brand: enQuireBio™ QP7274-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
hepatitis protein 1 | |
Research Use Only | |
X | |
Recombinant Protein | |
E. coli | |
MAARLCCQLD PARDVLCLRP VGAESRGRPF SGSLGTLSSP SPSAVPTDHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQILP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCAPAPCNF FTSA | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |