missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant hepatitis protein 1

A DNA sequence encoding the Hepatitis B virus genotype D subtype ayw (isolate France/Tiollais/1979) (HBV-D) Protein X, was expressed in the hosts and tags indicated. opt

Brand:  enQuireBio™ QP7274-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

hepatitis protein 1
Research Use Only
X
Recombinant Protein
E. coli
MAARLCCQLD PARDVLCLRP VGAESRGRPF SGSLGTLSSP SPSAVPTDHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQILP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCAPAPCNF FTSA
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
Hepatitis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions