missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp120 LAV Protein
A cDNA sequence encoding the HIV-1 gp120 LAV was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | HIV-1 gp120 LAV Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929818
|
enQuireBio™
QP12245-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919818
|
enQuireBio™
QP12245-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909818
|
enQuireBio™
QP12245-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-1 gp120 LAV Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
The protein solution contains 10mM Tris-Cl pH 7.6, 150mM NaCl and 0.01% Triton N-101. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
IPGEKLWVTV YYGVPVWKEA TTTLFCASDA KAYDTEVHNV ATHACVPTDP NPQEVVLVNV TENFNMWKND MVEQMHEDII SLWDQSLKPC VKLTPLCVSL KCTDLKNDTN TNSSSGRMIM EKGEIKNCSF NISTSIRDKV QKEYAFFYKL DIVPIDNTSY RLISCNTSVI TQACPKVSFE PIPIHYCAPA GFAILKCNNK TFNGTGPCTN VSTVQCTHGI RPVVSTQLLL NGSLAEEDVV IRSANFTDNA KTIIVQLNTS VEINCTRPNN NTRKSIRIQR GPGRAFVTIG KIGNMRQAHC NISRAKWNAT LKQIASKLRE QFGNNKTIIF KQSSGGDPEI VTHSFNCGGE FFYCNSTQLF NSTWFNSTWS TEGSNNTEGS DTITLPCRIK QFINMWQEVG KAMYAPPISG QIRCSSNITG LLLTRDGGNN NNGSEIFRPG GGDMRDNWRS ELYKYKVVKI EPLGVAPTKA KRRVVQREKR | |
Greater than 90.0% as determined by HPLC analysis and SDS-PAGE. |