missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp120 Nef Mosaic Protein
A cDNA sequence encoding the HIV-1 gp120 Nef Mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12247-500ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 gp120 Nef Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl, pH 4.5, 25mM Sodium Phosphate and 8M urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
His | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |