missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 p66 pol Protein
A cDNA sequence encoding the HIV-1 p66 pol was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | HIV-1 p66 pol Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949858
|
enQuireBio™
QP12259-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939858
|
enQuireBio™
QP12259-10UG |
10 μg |
288.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929858
|
enQuireBio™
QP12259-100UG |
100 μg |
2610.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-1 p66 pol Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by HPLC analysis & SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The protein solution contains 30mM Tris pH 7, 0.15M NaCl and 0.2mM EDTA. |