missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 Protease Protein
A cDNA sequence encoding the HIV-1 Protease was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | HIV-1 Protease Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979858
|
enQuireBio™
QP12260-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959858
|
enQuireBio™
QP12260-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969858
|
enQuireBio™
QP12260-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-1 Protease Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
The HIV-1 Protease filtered (0.4microm) solution is formulated in 20mM Tris, 20mM MES, 200mM NaCl, 1mM EDTA, 10% (v/v) glycerol and 0.05% 2-mercaptoethanol, pH 6.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF | |
Greater than 95.0% as determined by SDS-PAGE. |