missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 TAT Cys22 Protein
A cDNA sequence encoding the HIV-1 TAT Cys22 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12266-2ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 TAT Cys22 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized with 0.1% glycerol. |