missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 Envelope Protein
A cDNA sequence encoding the HIV-2 Envelope was constructed and used to recombinantly synthesize the protein.
228.00 € - 1450.00 €
Specifications
Name | HIV-2 Envelope Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969878
|
enQuireBio™
QP12267-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989878
|
enQuireBio™
QP12267-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979878
|
enQuireBio™
QP12267-1MG |
1 mg |
1450.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-2 Envelope Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl and 6M urea, pH 7.2. |