missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 gp46 mosaic Protein
A cDNA sequence encoding the HTLV-1 gp46 mosaic was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | HIV HTLV-1 gp46 mosaic Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990149
|
enQuireBio™
QP12355-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910159
|
enQuireBio™
QP12355-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900159
|
enQuireBio™
QP12355-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV HTLV-1 gp46 mosaic Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
HTLV-1 gp46 protein is >95% pure as determined by 10% SDS-PAGE (coomassie staining) and RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM NaPO4 pH 6.0, 0.1% SDS, 1mM DTT, 1mM EDTA. |