missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV NS3 His Protein
A cDNA sequence encoding the HCV NS3 His was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12175-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSV HCV NS3 His Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSVAHL HAPTGSGKST KVPAAYAAQG YKVLVLNPSV AATLGFGAYM SKAHGVDPNI RTGVRTITTG SPITYSTYGK FLADGGCSGG AYDIIICDEC HSTDATSILG IGTVLDQAET AGARLVVLAT ATPPGSVTVS HPNIEEVALS TTGEIPFYGK AIPLEVIKGG RHLIFCHSKK KCDELAAKLV ALGINAVAYY RGLDVSVIPT SGDVVVVSTD ALMTGFTGDF DSVIDCNT | |
Protein is >95% pure as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
HCV NS3 in 20mM Tris-HCl pH 8.0, 1mM DTT and 10% Glycerol. |