missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human A-kinase anchor protein 1
A DNA sequence encoding the Homo sapiens (Human) A-kinase anchor protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7671-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003877.2 | |
A-kinase anchor protein 1 | |
Research Use Only | |
AKAP4 | |
Recombinant Protein | |
E. coli | |
QSPSAPPAKP PSTQRAVISP DGECSIDDLS FYVNRLSSLV IQMAHKEIKE KLEGKSKCLH HSICPSPGNK ERISPRTPAS KIASEMAYEA VELTAAEMRG TGEESREGGQ KSFLYSELSN KSKSGDKQMS QRESKEFADS ISKGLMVYAN QVASDMMVSL MKTLKVHSSG KPIPASVVLK RVLLRHTKEI VSDLIDSCMK NLHNITGVLM TDSDFVSAVK RNLFNQWKQN ATDIMEAMLK RLVSALIGEE KETKSQSLSY ASLKAGSHDP KCRNQSLEFS TMKAEMKERD KGKMKSDPCK SLTSAEKVGE HILKEGLTIW NQKQGNSCKV ATKACSNKDE KGEKINASTD SLAKDLIVSA LKLIQYHLTQ QTKGKDTCEE DCPGSTMGYM AQSTQYEKCG GGQSAKALSV KQLESHRAPG PSTCQKENQH LDSQKMDMSN IVLMLIQKLL NENPFKCEDP CEGENKCSEP RASKAASMSN RSDKAEEQCQ EHQELDCTSG MKQANGQFID KLVESVMKLC LIMAKYSNDG AALAELEEQA ASANKPNFRG TRCIHSGAMP QNYQDSLGHE VIVNNQCSTN SLQKQLQAVL QWIAASQFNV PMLYFMGDKD GQLEKLPQVS AKAAEKGYSV GGLLQEVMKF AKERQPDEAV GKVARKQLLD WLLANL | |
Greater than 80% as determined by SDS-PAGE. |
8852 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |