missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACADM Protein
A DNA sequence encoding the Homo sapiens (Human) ACADM, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_000007.1 |
---|---|
Gene ID (Entrez) | 34 |
Name | ACADM Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956389
|
enQuireBio™
QP5605-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976389
|
enQuireBio™
QP5605-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966389
|
enQuireBio™
QP5605-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15986389
|
enQuireBio™
QP5605-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15996389
|
enQuireBio™
QP5605-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906399
|
enQuireBio™
QP5605-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000007.1 | |
ACADM Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
34 | |
Research Use Only | |
ACADM | |
Recombinant Protein | |
E. coli | |
KANRQREPGL GFSFEFTEQQ KEFQATARKF AREEIIPVAA EYDKTGEYPV PLIRRAWELG LMNTHIPENC GGLGLGTFDA CLISEELAYG CTGVQTAIEG NSLGQMPIII AGNDQQKKKY LGRMTEEPLM CAYCVTEPGA GSDVAGIKTK AEKKGDEYII NGQKMWITNG GKANWYFLLA RSDPDPKAPA NKAFTGFIVE ADTPGIQIGR KELNMGQRCS DTRGIVFEDV KVPKENVLIG DGAGFKVAMG AFDKTRPVVA AGAVGLAQRA LDEATKYALE RKTFGKLLVE HQAISFMLAE MAMKVELARM SYQRAAWEVD SGRRNTYYAS IAKAFAGDIA NQLATDAVQI LGGNGFNTEY PVEKLMRDAK IYQIYEGTSQ IQRLIVAREH IDKYKN | |
Greater than 90% as determined by SDS-PAGE. |