missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACAT1 Protein

A cDNA sequence encoding the ACAT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10936-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985618

  • 3401.86 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ACAT1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ACAT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl, 10% glycerol and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSVSKPTLK EVVIVSATRT PIGSFLGSLS LLPATKLGSI AIQGAIEKAG IPKEEVKEAY MGNVLQGGEG QAPTRQAVLG AGLPISTPCT TINKVCASGM KAIMMASQSL MCGHQDVMVA GGMESMSNVP YVMNRGSTPY GGVKLEDLIV KDGLTDVYNKIHMGSCAENT AKKLNIARNE QDAYAINSYT RSKAAWEAGK FGNEVIPVTV TVKGQPDVVV KEDEEYKRVD FSKVPKLKTV FQKENGTVTA ANASTLNDGA AALVLMTADA AKRLNVTPLA RIVAFADAAV EPIDFPIAPV YAASMVLKDV GLKKEDIAMW EVNEAFSLVV LANIKMLEIDPQKVNINGGA VSLGHPIGMS GARIVGHLTH ALKQGEYGLA SICNGGGGAS AMLIQKL
Videos
SDS
Documents
Certificates