missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACOT13 Protein
A cDNA sequence encoding the ACOT13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10942-2ug
Additional Details : Weight : 0.01000kg
Specifications
55856 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTSMTQSLRE VIKAMTKARN FERVLGKITL VSAAPGKVIC EMKVEEEHTN AIGTLHGGLT ATLVDNISTM ALLCTERGAP GVSVDMNITY MSPAKLGEDI VITAHVLKQG KTLAFTSVDL TNKATGKLIA QGRHTKHLGN | |
Greater than 95.0% as determined by SDS-PAGE. |
ACOT13 Protein | |
Research Use Only | |
ACOT13 | |
Human | |
His | |
The ACOT13 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |