missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACP1 / LMW-PTP Protein
A DNA sequence encoding the Homo sapiens (Human) ACP1 / LMW-PTP, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004291.1 |
---|---|
Gene ID (Entrez) | 52 |
Name | ACP1 / LMW-PTP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936409
|
enQuireBio™
QP5608-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956409
|
enQuireBio™
QP5608-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946409
|
enQuireBio™
QP5608-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966409
|
enQuireBio™
QP5608-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15976409
|
enQuireBio™
QP5608-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15986409
|
enQuireBio™
QP5608-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004291.1 | |
ACP1 / LMW-PTP Protein | |
Not determined. | |
Full Length of Isoform 1 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
52 | |
Research Use Only | |
ACP1 | |
Recombinant Protein | |
E. coli | |
MAEQATKSVL FVCLGNICRS PIAEAVFRKL VTDQNISENW RVDSAATSGY EIGNPPDYRG QSCMKRHGIP MSHVARQITK EDFATFDYIL CMDESNLRDL NRKSNQVKTC KAKIELLGSY DPQKQLIIED PYYGNDSDFE TVYQQCVRCC RAFLEKAH | |
Greater than 80% as determined by SDS-PAGE. |