missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACSF2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ACSF2, mitochondrial, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001275897.1 |
---|---|
Gene ID (Entrez) | 80221 |
Name | ACSF2, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16021981
|
enQuireBio™
QP7983-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041981
|
enQuireBio™
QP7983-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031981
|
enQuireBio™
QP7983-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051981
|
enQuireBio™
QP7983-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061981
|
enQuireBio™
QP7983-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071981
|
enQuireBio™
QP7983-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001275897.1 | |
ACSF2, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
80221 | |
Research Use Only | |
ACSF2 | |
Recombinant Protein | |
E. coli | |
LSSREVDRMV STPIGGLSYV QGCTKKHLNS KTVGQCLETT AQRVPEREAL VVLHEDVRLT FAQLKEEVDK AASGLLSIGL CKGDRLGMWG PNSYAWVLMQ LATAQAGIIL VSVNPAYQAM ELEYVLKKVG CKALVFPKQF KTQQYYNVLK QICPEVENAQ PGALKSQRLP DLTTVISVDA PLPGTLLLDE VVAAGSTRQH LDQLQYNQQF LSCHDPINIQ FTSGTTGSPK GATLSHYNIV NNSNILGERL KLHEKTPEQL RMILPNPLYH CLGSVAGTMM CLMYGATLIL ASPIFNGKKA LEAISRERGT FLYGTPTMFV DILNQPDFSS YDISTMCGGV IAGSPAPPEL IRAIINKINM KDLVVAYGTT ENSPVTFAHF PEDTVEQKAE SVGRIMPHTE ARIMNMEAGT LAKLNTPGEL CIRGYCVMLG YWGEPQKTEE AVDQDKWYWT GDVATMNEQG FCKIVGRSKD MIIRGGENIY PAELEDFFHT HPKVQEVQVV GVKDDRMGEE ICACIRLKDG EETTVEEIKA FCKGKISHFK IPKYIVFVTN YPLTISGKIQ KFKLREQMER HLNL | |
Greater than 90% as determined by SDS-PAGE. |